Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 0.1 M (0.1 N) in methanol ≥99% volumetric solution, Fluka™
Potassium hydroxide 0.1 M (0.1 N) in methanol ≥99% volumetric solution, Fluka™
Catalog # 319333-2L
CAS Number:  
Potassium hydroxide 0.1 M (0.1 N) in methanol ≥99% volumetric solution, Fluka™
Catalog # 319333-2L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

Specification Test Results

  • Assay:
    0.0950 - 0.1050 N
  • Carbonate (as K₂CO₃):
    Max. 0.003 N
  • Traceable to NIST SRM:
    complying

Specification Test Results

About this item

 :
 :56.11 g/mol
Density:0.81 g/cm³
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:215-181-3
UN:1230
ADR:3,II
 :GHS08
 :GHS02
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553