Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide standardised solution 0.1 N in methanol
Potassium hydroxide standardised solution 0.1 N in methanol
Catalog # 42734.K2
CAS Number:  
Potassium hydroxide standardised solution 0.1 N in methanol
Catalog # 42734.K2
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1992
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
 :GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553