Order Entry
Austria
ContactUsLinkComponent
 
Potassium hydroxide 0.1 mol/l (0.1 N), AVS TITRINORM volumetric solution
Catalog # 31780.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydroxide 0.1 mol/l (0.1 N), AVS TITRINORM volumetric solution
Catalog # 31780.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • NIST traceable:
    Confirmed
  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l

Specification Test Results

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,III
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553