

Hydrobromic acid 47%, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323
:80.91 g/mol
Density:1.47 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-113-0
UN:1788
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-11 °C
- Boiling Point:126 °C (1013 hPa)
- Vapor Pressure:10.6 hPa (20 °C)
- Density:1.49 g/cm3 (20 °C)
- pH:(H2O, 20 °C) strongly acid
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Merck Ultrapur and Suprapur® High Purity Acids and Bases
Product Literature
msds_100306
SDS
ProductPage_100306
Product specification
Frequently Bought Together