
Potassium permanganate 0.1 M KMnO4 (0.5N) volumetric solution, Fluka™
:7722-64-7
:
:KMnO₄
:MFCD00011364
:158.03 g/mol
:100 °C (1.013 hPa)
:Ambient
:231-760-3
:9,III
:GHS07
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Frequently Bought Together