Order Entry
Austria
ContactUsLinkComponent
Acrylamide 30% in aqueous solution
Toxic
Health
Acrylamide 30% in aqueous solution
Catalog # ICNA11ACRL4001
Supplier:  MP Biomedicals
CAS Number:  
undefined
Toxic
Health
Acrylamide 30% in aqueous solution
Catalog # ICNA11ACRL4001
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :71.08 g/mol
EINECS:201-173-7
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:79-06-1
Formula:C₃H₅NO
MDL Number:MFCD00008032