Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 5 mol/l (10 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid 5 mol/l (10 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 191665V
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphuric acid 5 mol/l (10 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 191665V
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, Safebreak®
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    4.99 - 5.01 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589