
Sulphuric acid 2.5 mol/l (5 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30138.293
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):2.495 - 2.505 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
:98.08 g/mol
Density:1.146 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589