Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 0.25 mol/l (0.5 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sulphuric acid 0.25 mol/l (0.5 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7110.1000
Supplier:  Avantor
CAS Number:  
Sulphuric acid 0.25 mol/l (0.5 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7110.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (N):
    0.4975 - 0.5025
  • Chloride (Cl):
    max. 1
  • Iron (Fe):
    max. 0.5
  • Heavy Metals (as Pb):
    max. 0.3
  • Phosphate (PO4):
    max. 1

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589