Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution
Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution
Catalog # MC302865.6010
Supplier:  PROLABO CMP
CAS Number:  
Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution
Catalog # MC302865.6010
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589