Hydrochloric acid 4 mol/l (4 N), VOLUSOL® volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
:GHS07
:GHS05
Specification Test Results
- Titer:3.984 - 4.016 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC310701.1000 spec_VWRC310701.1000 spec_VWRC310701.5000
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together