Order Entry
Austria
ContactUsLinkComponent
Guanidinium hydrochloride solution 6 M, GenAR® for biotechnology, Macron Fine Chemicals™
Irritant
Guanidinium hydrochloride solution 6 M, GenAR® for biotechnology, Macron Fine Chemicals™
Certificates
undefined
Irritant
Guanidinium hydrochloride solution 6 M, GenAR® for biotechnology, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Appearance (clear, colorless liquid):
    Passes Test
  • Cyanide (CN):
    Passes Test
  • Heavy Metals (as Pb):
    ≤10 ppm
  • Identification (by FTIR):
    Passes Test
  • ACS - Iron (Fe):
    ≤3 ppm
  • Molarity (by titrimetry):
    5.8 - 6.2
  • pH (1:10 dilution):
    4.5 - 6.5
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.1

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together