EDTA disodium salt 0.2 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
EDTA disodium salt 0.2 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
:139-33-3
:
:C₁₀H₁₄N₂Na₂O₈
:MFCD00070672
:336.21 g/mol
:Ambient
:382-266-9
- Factor:0.999 - 1.001
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals