



Glutaraldehyde 8% in aqueous solution
Artikelnr. ACRO410971000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:Clear solution
- Color scale:≤50 APHA
- Specific gravity:(20/20°C) 0.918 to 1.123
- Titration with NaOH:7 to 9 % v/v
Specification Test Results
About this item
:
:100.12 g/mol
Storage Temperature:Refrigerator
EINECS:203-856-5
UN:3265
ADR:8,II
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025