Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt standardised solution 0.1 M (0.200 N) in aqueous solution
EDTA disodium salt standardised solution 0.1 M (0.200 N) in aqueous solution
Catalog # 39783.K2
CAS Number:  
EDTA disodium salt standardised solution 0.1 M (0.200 N) in aqueous solution
Catalog # 39783.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672