EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.0499 - 0.0501 mol/l
- NIST traceable:Confirmed
Specification Test Results
Frequently Bought Together