Order Entry
Austria
ContactUsLinkComponent
Glutaraldehyde 50% in aqueous solution for synthesis, Sigma-Aldrich®
Glutaraldehyde 50% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Glutaraldehyde 50% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -33 °C
  • Boiling Point:
    100.7 °C (1013 hPa)
  • Vapor Pressure:
    22 hPa (20 °C)
  • Flash Point:
    95 °C
  • Density:
    1.13 g/cm3 (20 °C)
  • pH:
    3.1 - 4.5 (H2O, 20 °C)
  • Ignition Point:
    395 °C
  • Viscosity kinematic:
    12.75 mm2/s (25 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together