Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt 0.01 mol/l (0.02 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
EDTA disodium salt 0.01 mol/l (0.02 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7613.1000
Supplier:  Avantor
CAS Number:  
EDTA disodium salt 0.01 mol/l (0.02 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7613.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Molarity:
    0.0099 - 0.0101

Specification Test Results

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672