Order Entry
Austria
ContactUsLinkComponent
Glutaraldehyde 50% in aqueous solution, Reagent Grade
Toxic
Corrosive
Health
Environment
Glutaraldehyde 50% in aqueous solution, Reagent Grade
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Corrosive
Health
Environment
Glutaraldehyde 50% in aqueous solution, Reagent Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Boiling Range:
    >95 °C
  • Color (APHA):
    ≤100
  • Glutaraldehyde:
    50 – 51.5%
  • Methanol:
    ≤1%
  • pH at 25 °C:
    ≤5
  • Solidification range:
    ≥–20 °C
  • Specific gravity:
    1.125 – 1.135

Specification Test Results

Related Information

Frequently Bought Together