Order Entry
Austria
ContactUsLinkComponent
Oxalic acid 0.1 M (COOH)2 (0.2N) ≥98% volumetric solution, Fluka™
Oxalic acid 0.1 M (COOH)2 (0.2N) ≥98% volumetric solution, Fluka™
Catalog # 34287-1L
CAS Number:  
Oxalic acid 0.1 M (COOH)2 (0.2N) ≥98% volumetric solution, Fluka™
Catalog # 34287-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :90.04 g/mol
Storage Temperature:Ambient
EINECS:231-791-2
ADR:8,III
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573