Order Entry
Austria
ContactUsLinkComponent
Oxalic acid 0.05 mol/l (0.1 N), AVS TITRINORM volumetric solution
Oxalic acid 0.05 mol/l (0.1 N), AVS TITRINORM volumetric solution
Catalog # 30093.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxalic acid 0.05 mol/l (0.1 N), AVS TITRINORM volumetric solution
Catalog # 30093.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0499 - 0.0501 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :90.04 g/mol
Storage Temperature:Ambient
EINECS:205-634-3
ADR:8,III
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573