Order Entry
Austria
ContactUsLinkComponent
Oxalic acid concentrate for dilution to 0.05 mol/l (0.1 N), Titrisol®, Supelco®
Corrosive
Oxalic acid concentrate for dilution to 0.05 mol/l (0.1 N), Titrisol®, Supelco®
Catalog # 1.09965.0001
Supplier:  Merck
CAS Number:  
Corrosive
Oxalic acid concentrate for dilution to 0.05 mol/l (0.1 N), Titrisol®, Supelco®
Catalog # 1.09965.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.02 g/cm3 (20 °C)
  • pH:
    1 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :90.04 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ100
EINECS:205-634-3
 :GHS05
Storage Temperature:Ambient
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573