ContactUsLinkComponent
Oxalic acid concentrate for dilution to 0.05 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Oxalic acid concentrate for dilution to 0.05 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4665.
 :  Avantor
 :  
Oxalic acid concentrate for dilution to 0.05 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4665.
 :  Avantor
 :  
  • Pk:
    210 mL
  • Seq:
    2545
  • Titer (N):
    0.0998-0.1002
 :
 :90.04 g/mol
 :8,III
 :Dose for preparation of 1 litre of solution.
 :205-634-3
 :GHS07
 :Ambient
 :144-62-7
 :C₂H₂O₄
 :MFCD00002573