Oxalic acid concentrate for dilution to 0.005 mol/l (0.01 N), Titrisol®, Supelco®
Catalog # 1.09932.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Seq:0501
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.002 g/cm3 (20 °C)
- pH:1.5 (H2O, 20 °C)
Specification Test Results
About this item
:
:90.04 g/mol
Storage Temperature:Ambient
ADR:8,III
:M-Clarity™ quality level = MQ100
EINECS:205-634-3
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573