Order Entry
Austria
ContactUsLinkComponent
Glutaraldehyde 25% in aqueous solution for electron microscopy, acc. to P.J. Anderson, Sigma-Aldrich®
Corrosive
Health
Irritant
Environment
Glutaraldehyde 25% in aqueous solution for electron microscopy, acc. to P.J. Anderson, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Health
Irritant
Environment
Glutaraldehyde 25% in aqueous solution for electron microscopy, acc. to P.J. Anderson, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (alkalimetric):
    24.7 - 25.7 %
  • Identity:
    conforms
  • Free acid (as glutaric acid):
    ≤ 0,2 %
  • Methanol (HS-GC):
    ≤ 0,3 %
  • Optical purity (E 280 nm / E 235 nm):
    ≥ 3,0
  • pH:
    >3,0 (H₂O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together