



Glutaraldehyde 25% in water for electron microscopy, purified
:111-30-8
:
:C₅H₈O₂
:MFCD00007025
:100.12 g/mol
:101 °C (1013 hPa)
:–5 °C
:1.06 g/cm³
:≥ 71 °C
:203-856-5
:3265
:8,II
:GHS05
:GHS07
:GHS08
:GHS09
:Refrigerator
- Appearance:Clear to slightly hazy solution
- Assay:25 to 27 % w/w
- Color scale:≤100 APHA
- Specific gravity:(20/20°C) 1.000 to 1.133
- pH:3.1 to 4.5 (at 25°C)
Frequently Bought Together