(±)-1-Phenyl-1-propanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Phenyl-1-propanol, (±)-α-Ethylbenzyl alcohol
CAS Number:93-54-9
:
Formula:C₆H₅CH(OH)CH₂CH₃
MDL Number:MFCD00004564
Merck Index:14,3768
Boiling Pt:217…219 °C
Density:0.994
Flash Pt:95 °C (203 °F)
:136.19 g/mol
EINECS:202-256-0
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together