DL-Tyrosine
Catalog # EHERC17896000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:181.19 g/mol
Merck Index:12,09970
Boiling Pt:378 °C (1013 hPa)
Melting Pt:285 °C
EINECS:209-113-1
CAS Number:556-03-6
Formula:C₉H₁₁NO₃
MDL Number:MFCD00063074