
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-41-5
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00004547
:118.18 g/mol
Merck Index:13,04730
Boiling Pt:198 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.923 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:93 °C
EINECS:203-489-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together