Order Entry
Austria
ContactUsLinkComponent
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -40 °C
  • Molar Mass:
    118.17 g/mol
  • Boiling Point:
    196 °C
  • Vapor Pressure:
    0.07 hPa (20 °C)
  • Flash Point:
    93 °C DIN 51758
  • Refractive Index:
    1.427 (20 °C, 589 nm)
  • Explosion Limit:
    1 - 9.9 %(V)
  • Density:
    0.92 g/cm3 (20 °C)
  • Ignition Point:
    425 °C DIN 51794
  • Synonyms:
    Hexylene glycol
  • Hill Formula:
    C6H14O2

Specification Test Results

Related Information

Frequently Bought Together