Order Entry
Austria
ContactUsLinkComponent
(±)-2-Methyl-2,4-pentanediol 99%
(±)-2-Methyl-2,4-pentanediol 99%
Certificates
undefined
(±)-2-Methyl-2,4-pentanediol 99%
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4260 to 1.4280 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤15 APHA

Specification Test Results

Related Information

Frequently Bought Together