Order Entry
Austria
ContactUsLinkComponent
(±)-4-Methyl-2-pentanol
Flammable
Irritant
(±)-4-Methyl-2-pentanol
Catalog # EHERC15122100
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Irritant
(±)-4-Methyl-2-pentanol
Catalog # EHERC15122100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :102.18 g/mol
Boiling Pt:132 °C (1013 hPa)
Melting Pt:–90 °C
Density:0.8093 g/cm³ (20 °C)
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:203-551-7
UN:2053
ADR:3,III
 :GHS02
 :GHS07
CAS Number:108-11-2
Formula:C₆H₁₄O
MDL Number:MFCD00004550