(±)-2-Octanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
CAS Number:123-96-6
:
Formula:C₈H₁₈O
MDL Number:MFCD00004591
:130.23 g/mol
Merck Index:13,06785
Boiling Pt:180 °C (1013 hPa)
Melting Pt:–39 °C
Density:0.818 g/cm³ (20 °C)
EINECS:204-667-0
UN:1987
ADR:3,III
:M-Clarity™ quality level = MQ300
Flash Pt:55 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together