ContactUsLinkComponent
Labels, hazard warning, floor marking
  BRDY223663
 :  Brady
undefined
Labels, hazard warning, floor marking
  BRDY223663
 :  Brady
  • Colour:
    Black on yellow
  • Description:
    Hazard floor labels
  • Dimensions:
    500×500 mm
  • Material:
    Polyester
  • Seq:
    07
  • Model:
    PIC 306-TRI 500-B7538
  • Pk:
    1
These floor marking labels are made of durable vinyl and are provided with a non-slip laminate layer. They are used to warn people for potential hazards.
  • Resistant to abrasion
  • Passable