

DL-Malic acid 99-100.5%, high purity
Catalog # 0664-2.5KG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:HDPE bottle
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Fumaric Acid:<1%
- Heavy Metals (as Pb):≤0.001%
- Insolubles:≤0.1%
- Maleic Acid:≤0.05%
- Purity:99 – 100.5%
- Residue on Ignition:≤0.1%
Specification Test Results
About this item
:
:134.09 g/mol
Merck Index:13,05730
Boiling Pt:306 °C (1013 hPa)
Melting Pt:131…132 °C
Density:1.601 g/cm³ (20 °C)
EINECS:210-514-9
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:617-48-1
Formula:HOOCCH(OH)CH₂COOH
MDL Number:MFCD00064212