Order Entry
Austria
ContactUsLinkComponent
 
Flammable
Irritant
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8033.1000
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8033.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (by GC) (exclusive of preservative):
    min. 98.0 %
  • Carbonyl Compounds (as HCHO):
    max. 0.001 %
  • Color (APHA):
    max. 10
  • Peroxide (as H2O2):
    max. 1 ppm
  • Preservative (BHT):
    min. 7 ppm
  • Preservative (C2H5OH):
    1.5 - 3.5 %
  • Preservative (H2O):
    max. 0.5 %
  • Residue after Evaporation:
    max. 0.001 %
  • Titrable Acid (µeq/g):
    max. 0.2

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
REACH:01-2119535785-29
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646