

Diisopropyl ether ≥99% stabilised, TECHNICAL
Catalog # 24896.291
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 99 %
- Appearance:Clear colourless liquid
- IR Spectrum:Passes test
- Peroxides:Not detectable
- Propan-2-ol:Max. 1.5 %
Specification Test Results
About this item
:
:102.18 g/mol
Merck Index:13,05232
Boiling Pt:68.5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.726 g/cm³ (20 °C)
Flash Pt:–28 °C
EINECS:203-560-6
UN:1159
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 10 ppm
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:108-20-3
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880