Order Entry
Austria
ContactUsLinkComponent
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Catalog # MOLE18945167
Supplier:  Molekula
CAS Number:  
Tween® 20 (Polysorbate)
Catalog # MOLE18945167
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :1227.53 g/mol
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986