Order Entry
Austria
ContactUsLinkComponent
di-Sodium L(+)-tartrate dihydrate ≥99%, GPR RECTAPUR®
di-Sodium L(+)-tartrate dihydrate ≥99%, GPR RECTAPUR®
Catalog # 28174.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
di-Sodium L(+)-tartrate dihydrate ≥99%, GPR RECTAPUR®
Catalog # 28174.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99 %
  • Loss on drying (150°C):
    Max. 17.0 %
  • C2O4 (Oxalate):
    Max. 0.05 %
  • Cl (Chloride):
    Max. 50 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 100 ppm
  • Pb (Lead):
    Max. 10 ppm
  • Zn (Zinc):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :230.08 g/mol
Merck Index:12,08833
Melting Pt:57 °C
Density:1.818 g/cm³ (20 °C)
EINECS:212-773-3
CAS Number:6106-24-7
Formula:NaOOC(CHOH)₂COONa·2H₂O
MDL Number:MFCD00150035