Order Entry
Austria
ContactUsLinkComponent
di-Sodium L(+)-tartrate dihydrate, EMSURE® for analysis, Supelco®
di-Sodium L(+)-tartrate dihydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
di-Sodium L(+)-tartrate dihydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    290 g/l (20 °C)
  • Melting Point:
    57 °C
  • Molar Mass:
    230.08 g/mol
  • Bulk Density:
    460 kg/m3
  • Density:
    1.82 g/cm3
  • pH:
    8 (50 g/l, H2O, 25 °C)

Specification Test Results

Related Information

Frequently Bought Together