Tween® 20 (Polysorbate) for molecular biology
Catalog # 437082Q
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellowish granules
- DNases:Not detected
- RNases:Not detected
- Proteases:Not detected
- Density (20/4):1.095 - 1.105
- Hydroxyl value:96 - 108
- Saponification value:40.0 - 50.0
Specification Test Results
About this item
:
:1227.53 g/mol
Density:1.1 g/cm³ (20 °C)
Flash Pt:149 °C (closed cup)
Storage Temperature:Ambient
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986