Order Entry
Austria
ContactUsLinkComponent
VWR® ExoCleanUp FAST, One-step PCR Clean-up
VWR® ExoCleanUp FAST, One-step PCR Clean-up
Supplier:  VWR Chemicals
Certificates
undefined
VWR® ExoCleanUp FAST, One-step PCR Clean-up
Supplier:  VWR Chemicals
Certificates

Specifications

  • Method / Format:
    Enzymatic clean-up
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Sample size:
    1 × 1 ml
  • Description:
    ExoCleanUp FAST PCR clean-up reagent

Specifications

Frequently Bought Together