Tween® 80 (Polysorbate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9005-65-6
:
MDL Number:MFCD00082107
Merck Index:13,07664
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:–21 °C
Density:1.062 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>149 °C
Storage Temperature:Ambient
EINECS:500-019-9
Frequently Bought Together