Order Entry
Austria
ContactUsLinkComponent
 
Tween® 80 (Polysorbate) for molecular biology, Sigma-Aldrich®
Catalog # SIALP5188-100ML
Supplier:  Merck
CAS Number:  
Tween® 80 (Polysorbate) for molecular biology, Sigma-Aldrich®
Catalog # SIALP5188-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Melting Pt:100 °C
 :M-Clarity™ quality level = MQ200
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:500-019-9
CAS Number:9005-65-6
MDL Number:MFCD00082107