Order Entry
Austria
ContactUsLinkComponent
L(+)-Asparagine monohydrate 99.0-101.0% (dry basis), EMPROVE® EXPERT Ph. Eur. suitable for use as excipient, SAFC®
L(+)-Asparagine monohydrate 99.0-101.0% (dry basis), EMPROVE® EXPERT Ph. Eur. suitable for use as excipient, SAFC®
Certificates
L(+)-Asparagine monohydrate 99.0-101.0% (dry basis), EMPROVE® EXPERT Ph. Eur. suitable for use as excipient, SAFC®
Certificates

Specification Test Results

  • Assay (perchloric acid titration, calculated on dried substance):
    99.0 - 101.0 %
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    white to almost white crystalline powder or colorless crystals
  • Appearance of solution (20 g/l, CO₂-free water):
    clear and colorless
  • Spec. rotation (α 20/D, 100 g/l, hydrochloric acid 3 mol/l, calc. on dried substance):
    +33.7 to +36.0
  • pH (20 g/l, CO₂-free water):
    4.0 - 6.0
  • Chloride (Cl):
    ≤ 200 ppm
  • Sulphate (SO₄):
    ≤ 200 ppm
  • Fe (Iron):
    ≤ 10 ppm
  • NH₄ (Ammonium):
    ≤ 0.1 %
  • Aspartic acid:
    ≤ 0.5 %
  • Ninhydrin - positive substances (TLC) (Glutamic acid):
    ≤ 0.5 %
  • Ninhydrin - positive substances (TLC) (any other impurity):
    ≤ 0.5 %
  • Ethanol (HS-GC):
    ≤ 5000 ppm
  • Other residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Loss on drying (130 °C, 3 h):
    10.5 - 12.5 %
  • Bacterial endotoxins:
    ≤ 3.0 I.U./g

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together