ContactUsLinkComponent
Polysorbate 80, GPR RECTAPUR®
Polysorbate 80, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Polysorbate 80, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Identification:
    Passes test
  • Density (20/4):
    1.070 - 1.090
  • Hydroxyl value:
    65 - 80
  • Saponification value:
    45 - 55
  • Acid value:
    Max 2 mg KOH/g
  • Water content:
    Max 3%