Order Entry
Austria
ContactUsLinkComponent
 
 
Tween® 80 (Polysorbate), liquid, Cell Culture Grade
  ICNA02194725.1
 :  
 
Tween® 80 (Polysorbate), liquid, Cell Culture Grade
  ICNA02194725.1
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Identity Test A:
    Passes
  • Identity Test B:
    Passes
  • Identity Test C:
    Passes
  • Water Content:
    ≤3.0%
  • Residue on Ignition:
    ≤0.25%
  • Specific Gravity:
    1.06-1.09
  • Viscosity:
    300-500cps
  • Saponification Value:
    45-55

 

 

 :
 :13,07664
 :100 °C (lit.)
 :–21 °C
 :1.08 g/mL
 :>149 °C
 :Ambient
 :500-019-9
 :GHS07
 :9005-65-6
 :MFCD00082107