Order Entry
Austria
ContactUsLinkComponent
Tween® 80 (Polysorbate), Proteomics Grade
Tween® 80 (Polysorbate), Proteomics Grade
Supplier:  VWR Chemicals
Certificates
undefined
Tween® 80 (Polysorbate), Proteomics Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • DNase:
    None
  • Heavy metals:
    <0.001%
  • Hydroxyl number:
    65 − 80
  • Identification:
    Pass
  • Moisture (KF):
    3.0%
  • Protease:
    None
  • Residue on ignition:
    0.25%
  • RNase:
    None
  • Specific gravity:
    1.06 − 1.09
  • Viscosity:
    300 − 500 centistokes

Specification Test Results

Related Information

Frequently Bought Together