Order Entry
Austria
ContactUsLinkComponent
Paraformaldehyde 90%, pure
Flammable
Irritant
Health
Corrosive
Paraformaldehyde 90%, pure
Certificates
undefined
Flammable
Irritant
Health
Corrosive
Paraformaldehyde 90%, pure

Specification Test Results

  • Appearance:
    White Powder or granules or prills
  • Infrared spectrum:
    Conforms
  • Assay:
    89.5 to 93.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together