VWR®, Taq Plus 2X master mixes
VWR® Taq Plus is an optimised format of Taq DNA polymerase master mix and, therefore, is a good alternative to Taq DNA polymerase and Taq DNA polymerase master mix.
- Ready to use 2X master mix for greater convenience
- Significantly reduced set-up time
- Increased specificity compared to Taq DNA polymerase master mix
- Reduced risk of contamination
Ensures increased specificity and improved PCR performance, even on difficult templates. These reagents are suitable for standard PCR applications, screening and high throughput testing.
This PCR master mix is composed of Taq DNA polymerase and an optimised buffer system, including dNTPs and magnesium chloride (1,5 mM).
- Application:Standard amplification
- Format:Mastermix incl. dNTPs
- High fidelity:No
- Hot start:No
- Storage conditions:– 20 °C (long term), + 4 °C (6 months)
- Amplicon size:≤ 4kb
- Processivity:Standard
- Exonuclease activity:5’→3’
- Product overhang:3'A
- Concentration:2x
- Description:Taq Plus 2x Master Mix 1.5mM MgCl2 5000 reactions
- Size:25 x 5 ml
- Units:5000
Frequently Bought Together